Assay Details
| Gene Symbol: SRPX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014467.2 | NP_055282.1 | 2 - 3 | 559 | 84 | |
| GenBank mRNA | AF060567.1 | - | 2 - 3 | 497 | 84 | |
| AF393649.1 | - | 2 - 3 | 418 | 84 | ||
| AK075462.1 | - | 2 - 3 | 507 | 84 | ||
| BC020733.1 | - | 2 - 3 | 334 | 4769946 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000469.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
single organismal cell-cell adhesion
regulation of phosphorylation
cell motility
positive regulation of synapse assembly
vocalization behavior
positive regulation of cell migration involved in sprouting angiogenesis
receptor binding
protein binding
hepatocyte growth factor binding
identical protein binding
angiogenesis
single organismal cell-cell adhesion
regulation of phosphorylation
cell motility
positive regulation of synapse assembly
vocalization behavior
positive regulation of cell migration involved in sprouting angiogenesis
receptor binding
protein binding
hepatocyte growth factor binding
identical protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006026.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006026.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006026.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006026.hg.1 | TC0X000469.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006027.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006027.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006027.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006027.hg.1 | TC0X000469.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006032.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006032.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006032.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006032.hg.1 | TC0X000469.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006029.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006029.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006029.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006029.hg.1 | TC0X000469.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006024.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006024.hg.1 | TC0X000469.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006030.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006030.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006030.hg.1 | TC0X000469.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006031.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006031.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006031.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X006031.hg.1 | TC0X000469.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0X00153758.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153758.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153758.hg.1 | TC0X00007919.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00153764.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153764.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153764.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153764.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153764.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153764.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153764.hg.1 | TC0X00007919.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00153762.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153762.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153762.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153762.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153762.hg.1 | TC0X00007919.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00153765.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153765.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153765.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153765.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153765.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153765.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153765.hg.1 | TC0X00007919.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007919.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153760.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153760.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153760.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153760.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153760.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153760.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153760.hg.1 | TC0X00007919.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00153761.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153761.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153761.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153761.hg.1 | ENST00000373004.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153761.hg.1 | ENST00000481988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00153761.hg.1 | NM_014467.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00153761.hg.1 | TC0X00007919.hg.1 | Clariom D TC |

SRPX2
Genomic Map