| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006039.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006039.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006039.hg.1 |
TC0X000469.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000481988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000481988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000481988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000481988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
ENST00000481988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000481988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000481988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000481988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000481988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
ENST00000481988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007919.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153772.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153772.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153772.hg.1 |
ENST00000373004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153772.hg.1 |
NM_014467.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153772.hg.1 |
TC0X00007919.hg.1 |
Clariom D TC |