Assay Details
| Gene Symbol: PCNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002592.2 | NP_002583.1 | 1 - 2 | 121 | 95 | |
| GenBank mRNA | AK300558.1 | - | 1 - 2 | 40 | 95 | |
| AK311014.1 | - | 1 - 2 | 40 | 95 | ||
| CB990423.1 | - | 1 - 2 | 148 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000592.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK300558.1,NM_002592.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
telomere maintenance via recombination
telomere maintenance
DNA strand elongation involved in DNA replication
leading strand elongation
transcription-coupled nucleotide-excision repair
nucleotide-excision repair, DNA incision, 5'-to lesion
nucleotide-excision repair, DNA gap filling
mismatch repair
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
heart development
cell proliferation
protein sumoylation
translesion synthesis
epithelial cell differentiation
replication fork processing
positive regulation of deoxyribonuclease activity
nucleotide-excision repair, DNA incision
response to lipid
cellular response to UV
error-prone translesion synthesis
DNA damage response, detection of DNA damage
positive regulation of DNA repair
positive regulation of DNA replication
response to cadmium ion
error-free translesion synthesis
mitotic telomere maintenance via semi-conservative replication
purine-specific mismatch base pair DNA N-glycosylase activity
chromatin binding
damaged DNA binding
protein binding
enzyme binding
estrogen receptor binding
DNA polymerase processivity factor activity
receptor tyrosine kinase binding
dinucleotide insertion or deletion binding
MutLalpha complex binding
histone acetyltransferase binding
identical protein binding
DNA polymerase binding
G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
telomere maintenance via recombination
telomere maintenance
DNA strand elongation involved in DNA replication
leading strand elongation
transcription-coupled nucleotide-excision repair
nucleotide-excision repair, DNA incision, 5'-to lesion
nucleotide-excision repair, DNA gap filling
mismatch repair
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
heart development
cell proliferation
protein sumoylation
translesion synthesis
epithelial cell differentiation
replication fork processing
positive regulation of deoxyribonuclease activity
nucleotide-excision repair, DNA incision
response to lipid
cellular response to UV
error-prone translesion synthesis
DNA damage response, detection of DNA damage
positive regulation of DNA repair
positive regulation of DNA replication
response to cadmium ion
error-free translesion synthesis
mitotic telomere maintenance via semi-conservative replication
purine-specific mismatch base pair DNA N-glycosylase activity
chromatin binding
damaged DNA binding
protein binding
enzyme binding
estrogen receptor binding
DNA polymerase processivity factor activity
receptor tyrosine kinase binding
dinucleotide insertion or deletion binding
MutLalpha complex binding
histone acetyltransferase binding
identical protein binding
DNA polymerase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010291.hg.1 | ENST00000532690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008657.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008657.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008657.hg.1 | TC20000592.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008652.hg.1 | ENST00000379143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008652.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008652.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008652.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008652.hg.1 | TC20000592.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000158375.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158375.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158375.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158375.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158375.hg.1 | TC2000008279.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000158375.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158375.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158375.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158375.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158381.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158381.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158381.hg.1 | TC2000008279.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000158381.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158381.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008279.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010291.hg.1 | ENST00000532690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158372.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158372.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158372.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158372.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158372.hg.1 | TC2000008279.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000158372.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158372.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158372.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158372.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158374.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158374.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158374.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158374.hg.1 | NM_182649.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158374.hg.1 | TC2000008279.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000158374.hg.1 | ENST00000379143.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158374.hg.1 | ENST00000379160.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158374.hg.1 | NM_002592.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158374.hg.1 | NM_182649.1 | Refseq |

PCNA
Genomic Map