Assay Details
| Gene Symbol: BRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004333.4 | NP_004324.2 | 9 - 10 | 1244 | 146 | |
| XM_005250045.2 | XP_005250102.1 | 9 - 10 | 1191 | 146 | ||
| GenBank mRNA | AK314346.1 | - | 9 - 10 | 1191 | 146 | |
| BC101757.1 | - | 9 - 10 | 1205 | 8069263 | 146 | |
| BC112079.1 | - | 9 - 10 | 1205 | 8327547 | 146 | |
| M95712.2 | - | 9 - 10 | 1244 | 146 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001927.hg.1
Set Membership: |
|

BRAF
Genomic Map