| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017461.hg.1 |
ENST00000534738.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017461.hg.1 |
NM_181536.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017461.hg.1 |
TC16001255.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1600183856.hg.1 |
ENST00000335106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600183856.hg.1 |
ENST00000534738.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600183856.hg.1 |
ENST00000620267.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600183856.hg.1 |
NM_181536.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600183856.hg.1 |
TC1600010801.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600183856.hg.1 |
ENST00000335106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600183856.hg.1 |
ENST00000534738.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600183856.hg.1 |
ENST00000620267.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600183856.hg.1 |
NM_181536.1 |
Refseq |