| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000424164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000424164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000424164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000424164.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
NM_001042693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
NM_001042693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
NM_001042693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
NM_001042693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
NM_001042693.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008326.hg.1 |
NM_001042693.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000401050.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000424164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000424164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000424164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000424164.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000440303.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
ENST00000517870.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
NM_001042693.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
NM_001042693.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
NM_001042693.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
NM_001042693.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
NM_001042693.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008326.hg.1 |
NM_001042693.2 |
Refseq |