Assay Details
| Gene Symbol: SELT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016275.3 | NP_057359.2 | 4 | 503 | 154 | |
| GenBank mRNA | AF131856.1 | - | 4 | 458 | 154 | |
| AF195141.1 | - | 4 | 461 | 154 | ||
| AK075006.1 | - | 4 | 476 | 154 | ||
| AK300315.1 | - | 4 | 491 | 154 | ||
| AY358095.1 | - | 4 | 467 | 154 | ||
| AY358096.1 | - | 4 | 471 | 154 | ||
| BC006012.1 | - | 4 | 494 | 4333083 | 154 | |
| BC009556.1 | - | 4 | 435 | 3899430 | 154 | |
| BC009611.1 | - | 4 | 455 | 4093231 | 154 | |
| BC026350.1 | - | 4 | 483 | 4796883 | 154 | |
| BC036738.1 | - | 4 | 454 | 5540243 | 154 | |
| BC071699.1 | - | 4 | 446 | 5548565 | 154 | |
| CR457154.1 | - | 3 | 245 | 154 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SELT
Genomic Map