Assay Details
| Gene Symbol: HSPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021979.3 | NP_068814.2 | 1 | 649 | 100 | |
| GenBank mRNA | BC001752.1 | - | 2 | 343 | 3358246 | 100 |
| BT009815.1 | - | 1 | 267 | 100 | ||
| CD300159.1 | - | 1 | 67 | 100 | ||
| CN356507.1 | - | 1 | 432 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000391.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to unfolded protein
male meiosis
male meiosis I
spermatogenesis
spermatid development
response to heat
response to cold
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G2/M transition of mitotic cell cycle
protein refolding
synaptonemal complex disassembly
negative regulation of inclusion body assembly
positive regulation of calcium-transporting ATPase activity
protein binding
ATP binding
enzyme binding
unfolded protein binding
glycolipid binding
response to unfolded protein
male meiosis
male meiosis I
spermatogenesis
spermatid development
response to heat
response to cold
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G2/M transition of mitotic cell cycle
protein refolding
synaptonemal complex disassembly
negative regulation of inclusion body assembly
positive regulation of calcium-transporting ATPase activity
protein binding
ATP binding
enzyme binding
unfolded protein binding
glycolipid binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004461.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004461.hg.1 | ENST00000545222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004461.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004461.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004461.hg.1 | TC14000391.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004465.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004465.hg.1 | ENST00000394709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004465.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004465.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004465.hg.1 | TC14000391.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004463.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004463.hg.1 | ENST00000394709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004463.hg.1 | ENST00000545222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004463.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004463.hg.1 | TC14000391.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004464.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004464.hg.1 | ENST00000394709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004464.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004464.hg.1 | TC14000391.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004459.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004459.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004459.hg.1 | TC14000391.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004466.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004466.hg.1 | ENST00000394709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004466.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004466.hg.1 | TC14000391.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004462.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004462.hg.1 | ENST00000394709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004462.hg.1 | ENST00000545222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004462.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004462.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004462.hg.1 | TC14000391.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400151984.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151984.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151984.hg.1 | TC1400007443.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400151990.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151990.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151990.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151990.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400151990.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151990.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151990.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151990.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400151990.hg.1 | TC1400007443.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400151991.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151991.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151991.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400151991.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151991.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151991.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400151991.hg.1 | TC1400007443.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400151993.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151993.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151993.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400151993.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151993.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151993.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400151993.hg.1 | TC1400007443.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400151986.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151986.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400151986.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151986.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400151986.hg.1 | TC1400007443.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007443.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400151988.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151988.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151988.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400151988.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151988.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151988.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400151988.hg.1 | TC1400007443.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400151992.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151992.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151992.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151992.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400151992.hg.1 | ENST00000247207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151992.hg.1 | ENST00000394709.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151992.hg.1 | ENST00000554883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400151992.hg.1 | NM_021979.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400151992.hg.1 | TC1400007443.hg.1 | Clariom D TC |

HSPA2
Genomic Map