| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000587589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000587589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000587589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000587589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000587589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009394.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014769.hg.1 |
ENST00000309324.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014769.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014769.hg.1 |
TC19001080.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900175120.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175120.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900175120.hg.1 |
TC1900009394.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900175120.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175120.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900175119.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175119.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175119.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900175119.hg.1 |
TC1900009394.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900175119.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175119.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175119.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
ENST00000587589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
TC1900009394.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
ENST00000587589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900175116.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000309324.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000585605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000587589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000587589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000587589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000587589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000587589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000587950.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000590075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
ENST00000590389.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_001308240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_205767.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009394.hg.1 |
NM_205767.2 |
Refseq |