| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000588434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000588434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
ENST00000588434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006852.hg.1 |
ENST00000245382.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006852.hg.1 |
ENST00000319977.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006852.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006852.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006852.hg.1 |
TC17000563.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006851.hg.1 |
ENST00000245382.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006851.hg.1 |
ENST00000253405.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006851.hg.1 |
ENST00000319977.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006851.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006851.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17006851.hg.1 |
TC17000563.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000588073.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000588434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000588434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
ENST00000588434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007982.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700158363.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700158363.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700158363.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700158363.hg.1 |
ENST00000588434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700158363.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700158363.hg.1 |
NM_024032.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700158363.hg.1 |
TC1700007982.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700158363.hg.1 |
ENST00000245382.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700158363.hg.1 |
ENST00000319977.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700158363.hg.1 |
ENST00000585683.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700158363.hg.1 |
ENST00000588434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700158363.hg.1 |
NM_001171251.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700158363.hg.1 |
NM_024032.4 |
Refseq |