Assay Details
| Gene Symbol: MTDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178812.3 | NP_848927.2 | 7 - 8 | 1468 | 70 | |
| XM_011517367.2 | XP_011515669.1 | 8 - 9 | 1765 | 70 | ||
| XM_011517368.2 | XP_011515670.1 | 7 - 8 | 1699 | 70 | ||
| XM_011517369.2 | XP_011515671.1 | 8 - 9 | 1765 | 70 | ||
| XM_011517370.2 | XP_011515672.1 | 7 - 8 | 1675 | 70 | ||
| XM_017013966.1 | XP_016869455.1 | 6 - 7 | 1609 | 70 | ||
| GenBank mRNA | AF411226.1 | - | 7 - 8 | 1359 | 70 | |
| AF501310.1 | - | 7 - 8 | 1399 | 70 | ||
| AK000745.1 | - | 7 - 8 | 854 | 70 | ||
| AY082966.1 | - | 7 - 8 | 1218 | 70 | ||
| AY974040.1 | - | 7 - 8 | 1210 | 70 | ||
| BC001815.1 | - | 8 - 9 | 1408 | 70 | ||
| BC009324.2 | - | 8 - 9 | 1323 | 70 | ||
| BC014977.1 | - | 7 - 8 | 1440 | 70 | ||
| BC045642.1 | - | 7 - 8 | 1468 | 5298467 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000594.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
positive regulation of autophagy
lipopolysaccharide-mediated signaling pathway
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of angiogenesis
positive regulation of NF-kappaB transcription factor activity
positive regulation of protein kinase B signaling
bicellular tight junction assembly
RNA polymerase II transcription factor binding
transcription coactivator activity
double-stranded RNA binding
protein binding
poly(A) RNA binding
NF-kappaB binding
negative regulation of transcription from RNA polymerase II promoter
positive regulation of autophagy
lipopolysaccharide-mediated signaling pathway
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of angiogenesis
positive regulation of NF-kappaB transcription factor activity
positive regulation of protein kinase B signaling
bicellular tight junction assembly
RNA polymerase II transcription factor binding
transcription coactivator activity
double-stranded RNA binding
protein binding
poly(A) RNA binding
NF-kappaB binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008233.hg.1 | ENST00000336273.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008233.hg.1 | ENST00000519934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008233.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008233.hg.1 | TC08000594.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157257.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157257.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157257.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157257.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157257.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157257.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157257.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157257.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157257.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157257.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157257.hg.1 | TC0800008314.hg.1 | Clariom D TC |

MTDH
Genomic Map