Assay Details
| Gene Symbol: SHOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000451.3 | NP_000442.1 | 5 - 6 | 1326 | 59 | |
| GenBank mRNA | U89331.1 | - | 5 - 6 | 1328 | 59 | |
| Y11536.1 | - | 4 - 5 | 726 | 59 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000003.hg.1, TC0Y000003.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000049.hg.1 | ENST00000381578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000049.hg.1 | ENST00000554971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000049.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000049.hg.1 | TC0X000003.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000034.hg.1 | ENST00000381578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000034.hg.1 | ENST00000554971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000034.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000034.hg.1 | TC0Y000003.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000035.hg.1 | ENST00000381578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000035.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0Y000035.hg.1 | TC0Y000003.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000050.hg.1 | ENST00000381578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000050.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X000050.hg.1 | TC0X000003.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0X00144129.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144129.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144129.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144129.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144129.hg.1 | TC0X00006443.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00006443.hg.1 | NM_006883.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000334060.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0Y00006440.hg.1 | ENSTR0000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144128.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144128.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144128.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144128.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144128.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144128.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144128.hg.1 | TC0X00006443.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00144127.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144127.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144127.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144127.hg.1 | ENST00000381578.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144127.hg.1 | ENST00000554971.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00144127.hg.1 | NM_000451.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00144127.hg.1 | TC0X00006443.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 148829 | - | - |

SHOX
Genomic Map