| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02020999.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02020999.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02020999.hg.1 |
TC02001344.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
ENST00000440903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
ENST00000440903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
ENST00000440903.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02021000.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02021000.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02021000.hg.1 |
TC02001344.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200178090.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200178090.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200178090.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200178090.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200178090.hg.1 |
TC0200010894.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200178091.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200178091.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200178091.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200178091.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200178091.hg.1 |
TC0200010894.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
ENST00000295226.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
ENST00000440903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
ENST00000440903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
ENST00000440903.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010894.hg.1 |
NM_153038.1 |
Refseq |