Assay Details
Gene Symbol: NOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014062.2 | NP_054781.1 | 7 | 857 | 141 | |
NR_074074.1 | 6 | 726 | 141 | |||
GenBank mRNA | AB026125.1 | - | 7 | 815 | 141 | |
AF190161.1 | - | 7 | 785 | 141 | ||
AK001028.1 | - | 7 | 812 | 141 | ||
AK314307.1 | - | 7 | 812 | 141 | ||
AY487344.1 | - | 7 | 831 | 141 | ||
BC000050.2 | - | 4 | 444 | 141 | ||
BC064630.1 | - | 7 | 800 | 5740890 | 141 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001226.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16016947.hg.1 | ENST00000268802.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16016947.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16016947.hg.1 | TC16001226.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600182919.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600182919.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600182919.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600182919.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600182919.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600182919.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600182919.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600182919.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600182919.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600182919.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600182919.hg.1 | TC1600010734.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000268802.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000561677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000562416.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000563055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000564620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | ENST00000569871.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NM_014062.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600010734.hg.1 | NR_074074.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 169620 | - | - |
