Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006874.hg.1 |
ENST00000239852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006874.hg.1 |
ENST00000379477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006874.hg.1 |
ENST00000379480.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006874.hg.1 |
ENST00000430347.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006874.hg.1 |
ENST00000486236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006874.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006874.hg.1 |
TC13000602.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000430347.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000430347.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000430347.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000430347.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000473492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000473492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000473492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000473492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000473492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000492827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000492827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000492827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000492827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000492827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000498245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000498245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000498245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
ENST00000498245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000430347.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000430347.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000430347.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000430347.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000473492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000473492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000473492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000473492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000473492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000492827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000492827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000492827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000492827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000492827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000498245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000498245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000498245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
ENST00000498245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008720.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
ENST00000239852.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
ENST00000379477.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
ENST00000379480.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
ENST00000452359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
ENST00000480434.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
ENST00000497679.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
NM_004294.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158851.hg.1 |
TC1300008720.hg.1 |
Clariom D TC |