Assay Details
| Gene Symbol: UHRF1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017754.3 | NP_060224.3 | 18 - 19 | 4215 | 66 | |
| GenBank mRNA | AB126777.1 | - | 18 - 19 | 4050 | 66 | |
| AK122918.1 | - | 4 - 5 | 1841 | 66 | ||
| AK127691.1 | - | 5 - 6 | 1011 | 66 | ||
| BC137120.1 | - | 18 - 19 | 4045 | 9021117 | 66 | |
| BC137127.1 | - | 18 - 19 | 4045 | 9021124 | 66 | |
| BX640641.1 | - | 19 - 20 | 4180 | 66 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | ENST00000516297.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000192788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000452449.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | ENST00000516297.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007771.hg.1 | NM_017754.3 | Refseq |

UHRF1BP1
Genomic Map