| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012923.hg.1 |
ENST00000375475.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012923.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012923.hg.1 |
ENST00000505358.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012923.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012923.hg.1 |
TC11001030.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012921.hg.1 |
ENST00000375475.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012921.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012921.hg.1 |
ENST00000505358.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012921.hg.1 |
ENST00000541074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012921.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012921.hg.1 |
TC11001030.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012920.hg.1 |
ENST00000375475.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012920.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012920.hg.1 |
ENST00000505358.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012920.hg.1 |
ENST00000541074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012920.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012920.hg.1 |
TC11001030.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012922.hg.1 |
ENST00000375475.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012922.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012922.hg.1 |
ENST00000505358.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012922.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012922.hg.1 |
TC11001030.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012916.hg.1 |
ENST00000375475.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012916.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012916.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012916.hg.1 |
TC11001030.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012918.hg.1 |
ENST00000375475.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012918.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012918.hg.1 |
ENST00000505358.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012918.hg.1 |
ENST00000541074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012918.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11012918.hg.1 |
TC11001030.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009122.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100167673.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100167673.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100167673.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100167673.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100167673.hg.1 |
TC1100009122.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100167672.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100167672.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100167672.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100167672.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100167672.hg.1 |
TC1100009122.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100167675.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100167675.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100167675.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100167675.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100167675.hg.1 |
TC1100009122.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100167674.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100167674.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100167674.hg.1 |
ENST00000389586.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100167674.hg.1 |
NM_182495.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100167674.hg.1 |
TC1100009122.hg.1 |
Clariom D TC |