| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000663.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000663.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000663.hg.1 |
TC0Y000054.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006642.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006642.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006642.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006642.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006642.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006642.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006642.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006642.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006642.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006642.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006642.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006642.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000664.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000664.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000664.hg.1 |
TC0Y000054.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000665.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000665.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0Y000665.hg.1 |
TC0Y000054.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0Y00006642.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006642.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006642.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006642.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006642.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006642.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0Y00006642.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0Y00006642.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0Y00006642.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0Y00006642.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0Y00006642.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0Y00006642.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00145322.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00145322.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00145322.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00145322.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00145322.hg.1 |
TC0Y00006642.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0Y00145324.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00145324.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00145324.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00145324.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00145324.hg.1 |
TC0Y00006642.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0Y00145323.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00145323.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00145323.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00145323.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00145323.hg.1 |
TC0Y00006642.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0Y00145321.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00145321.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00145321.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00145321.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00145321.hg.1 |
TC0Y00006642.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0Y00145325.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00145325.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00145325.hg.1 |
ENST00000284856.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0Y00145325.hg.1 |
NM_004202.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0Y00145325.hg.1 |
TC0Y00006642.hg.1 |
Clariom D TC |