Assay Details
| Gene Symbol: NNAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181689.2 | NP_859017.1 | 1 - 2 | 197 | 66 | |
| GenBank mRNA | AI879211.1 | - | 1 - 2 | 153 | 66 | |
| HY021161.1 | - | 1 - 2 | 198 | 66 | ||
| U25034.1 | - | 1 - 2 | 142 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000274.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20003896.hg.1 | ENST00000062104.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003896.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003896.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003896.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003896.hg.1 | TC20000274.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007288.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003899.hg.1 | ENST00000062104.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003899.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003899.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003899.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003899.hg.1 | TC20000274.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003897.hg.1 | ENST00000062104.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003897.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003897.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003897.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20003897.hg.1 | TC20000274.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000150353.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150353.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150353.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150353.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150353.hg.1 | TC2000007288.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150353.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150353.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150353.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150353.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007288.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150356.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150356.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150356.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150356.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150356.hg.1 | TC2000007288.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150356.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150356.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150356.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150356.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150352.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150352.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150352.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150352.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150352.hg.1 | TC2000007288.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150352.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150352.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150352.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150352.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150355.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150355.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150355.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150355.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150355.hg.1 | TC2000007288.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150355.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150355.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150355.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150355.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150351.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150351.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150351.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150351.hg.1 | NM_181689.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150351.hg.1 | TC2000007288.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150351.hg.1 | ENST00000062104.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150351.hg.1 | ENST00000346199.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150351.hg.1 | NM_005386.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150351.hg.1 | NM_181689.2 | Refseq |

NNAT
Genomic Map