| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019075.hg.1 |
ENST00000455699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019075.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019075.hg.1 |
TC10001511.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019073.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019073.hg.1 |
TC10001511.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019074.hg.1 |
ENST00000455699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019074.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019074.hg.1 |
TC10001511.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011385.hg.1 |
ENST00000449882.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011385.hg.1 |
ENST00000449882.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011385.hg.1 |
ENST00000449882.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011385.hg.1 |
ENST00000449882.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011385.hg.1 |
ENST00000455699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011385.hg.1 |
ENST00000455699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011385.hg.1 |
ENST00000449882.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011385.hg.1 |
ENST00000449882.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011385.hg.1 |
ENST00000449882.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011385.hg.1 |
ENST00000449882.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011385.hg.1 |
ENST00000455699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011385.hg.1 |
ENST00000455699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011385.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177374.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177374.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177374.hg.1 |
TC1000011385.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000177376.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177376.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177376.hg.1 |
TC1000011385.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000177377.hg.1 |
ENST00000455699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177377.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177377.hg.1 |
ENST00000455699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177377.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177377.hg.1 |
TC1000011385.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000177378.hg.1 |
ENST00000455699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177378.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177378.hg.1 |
ENST00000455699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177378.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177378.hg.1 |
TC1000011385.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000177375.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177375.hg.1 |
NR_026835.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177375.hg.1 |
TC1000011385.hg.1 |
Clariom D TC |