Assay Details
| Gene Symbol: ZNF799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CB988846.1 | - | 4 | 670 | 91 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10886322 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10886330 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501658 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17109801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501683 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10886335 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10886325 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501738 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10886327 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10886321 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10886334 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501668 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501724 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10886320 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17109848 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501674 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10886329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501679 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501669 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501736 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501722 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501678 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10886332 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17109792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501655 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501665 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17109810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501717 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31501672 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 108809 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 899816 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 374616 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 505137 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 213998 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 105246 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 57298 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 97421 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 357536 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 216505 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 133522 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 175310 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 66093 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P90079 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 12602 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 56078 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P20960739 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P90079 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P90079 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886337 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501716 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109810 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886319 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886321 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109815 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501679 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501736 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501670 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501730 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109801 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886325 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886326 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501727 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501722 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886331 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886336 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501717 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886324 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501667 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886330 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886332 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886327 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886328 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501683 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501678 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109803 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886333 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886329 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886320 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886323 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501685 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501724 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886335 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501737 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501694 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501657 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501655 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886334 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109800 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501665 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109853 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501658 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501669 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501672 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886318 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501715 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501668 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109791 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109792 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886322 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501738 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501677 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109848 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501675 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501674 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31501738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31501730 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31501727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31501737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31501724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31501736 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886336 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501667 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501736 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109803 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886337 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886327 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109810 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501669 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886321 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109848 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886334 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886330 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886319 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501716 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501670 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109853 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501657 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886324 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886329 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886323 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109792 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501683 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501737 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501679 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501655 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501677 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501674 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501738 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501715 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501730 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886331 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109815 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501722 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886320 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501658 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501724 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501727 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501678 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109801 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109800 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886322 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886332 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886335 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501685 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501675 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501694 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886326 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501672 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17109791 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501717 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886325 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886333 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501668 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886328 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10886318 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31501665 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31501736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31501738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31501730 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31501737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31501727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31501724 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P90079 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 30312 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P34443303 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41143835 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17109810 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10886326 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17109801 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10886330 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17109803 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10886333 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10886334 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17109792 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10886337 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10886331 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10886329 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10886323 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P41143835 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001080821_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8034392 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8034391 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8034390 | - | - |
| Human Genome U95 Set | 35588_PM_at | - | - |
| Human Genome U95 Set | 35588_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011079 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011077 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011077 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011079 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3247154 | BC009517.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3247154 | AK027660.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3247154 | AL832890.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3247154 | NM_001080821.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1659265 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247154 | BC009517.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247154 | AK027660.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247154 | AL832890.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247154 | NM_001080821.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247154 | BC009517.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247154 | AK027660.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247154 | AL832890.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247154 | NM_001080821.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247154 | BC009517.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247154 | AK027660.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247154 | AL832890.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247154 | NM_001080821.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1659265 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1659265 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1659265 | - | - |

ZNF799
Genomic Map