| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
ENST00000355016.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
ENST00000355016.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
ENST00000355016.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
ENST00000355016.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
ENST00000355016.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
ENST00000355016.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
ENST00000417637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
ENST00000417637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
ENST00000417637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
ENST00000598335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
ENST00000598335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
ENST00000598335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
NM_001164416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
NM_001164416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
NM_001164416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
NM_001164416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
NM_001164416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008026.hg.1 |
NM_001164416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
ENST00000355016.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
ENST00000355016.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
ENST00000355016.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
ENST00000355016.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
ENST00000355016.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
ENST00000355016.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
ENST00000417637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
ENST00000417637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
ENST00000417637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
ENST00000598335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
ENST00000598335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
ENST00000598335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
NM_001164416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
NM_001164416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
NM_001164416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
NM_001164416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
NM_001164416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008026.hg.1 |
NM_001164416.1 |
Refseq |