| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017082.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017082.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017082.hg.1 |
TC04001255.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017083.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017083.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017083.hg.1 |
TC04001255.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017085.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017085.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017085.hg.1 |
TC04001255.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017084.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017084.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017084.hg.1 |
TC04001255.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010898.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174106.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174106.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174106.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174106.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174106.hg.1 |
TC0400010898.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400174104.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174104.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174104.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174104.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174104.hg.1 |
TC0400010898.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400174105.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174105.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174105.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174105.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174105.hg.1 |
TC0400010898.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400174108.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174108.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174108.hg.1 |
ENST00000356291.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174108.hg.1 |
NM_207407.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174108.hg.1 |
TC0400010898.hg.1 |
Clariom D TC |