| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08005625.hg.1 |
ENST00000399598.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08005625.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08005625.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08005625.hg.1 |
TC08000402.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000520732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000520732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000520732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000520732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000520732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000520732.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000521796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000521796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000521796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000521796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000521796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013241.hg.1 |
ENST00000451057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013241.hg.1 |
ENST00000451057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000520732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000520732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000520732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000520732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000520732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000520732.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000521796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000521796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000521796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000521796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000521796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800007735.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800153224.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153224.hg.1 |
ENST00000521796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153224.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153224.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153224.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800153224.hg.1 |
ENST00000399598.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153224.hg.1 |
ENST00000521796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153224.hg.1 |
ENST00000522978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153224.hg.1 |
ENST00000523409.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800153224.hg.1 |
NM_001077619.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800153224.hg.1 |
TC0800007735.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700013241.hg.1 |
ENST00000451057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013241.hg.1 |
ENST00000451057.1 |
Ensembl |