| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009993.hg.1 |
ENST00000615682.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009993.hg.1 |
ENST00000615682.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009309.hg.1 |
ENST00000422917.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009309.hg.1 |
ENST00000422917.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009309.hg.1 |
ENST00000422917.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009309.hg.1 |
ENST00000422917.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009309.hg.1 |
ENST00000422917.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009309.hg.1 |
ENST00000422917.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009993.hg.1 |
ENST00000615682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009993.hg.1 |
ENST00000615682.1 |
Ensembl |