Assay Details
| Gene Symbol: LOC153684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002146.1 | - | 1 - 2 | 300 | 76 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000505541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000505541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000505541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000515108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000515108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000515108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | ENST00000515108.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | NR_015447.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | NR_015447.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | NR_015447.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | NR_015447.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007284.hg.1 | NR_015447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000399543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000451894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000505541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000505541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000505541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000508913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000515108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000515108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000515108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | ENST00000515108.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | NR_015447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | NR_015447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | NR_015447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | NR_015447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007284.hg.1 | NR_015447.1 | Refseq |

LOC153684
Genomic Map