Assay Details
| Gene Symbol: SNRPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003093.2 | NP_003084.1 | 6 | 150 | ||
| NR_029472.1 | 5 | 150 | ||||
| GenBank mRNA | AI356808.1 | - | 1 | 107 | 150 | |
| BU153661.1 | - | 5 | 150 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000507.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06005312.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005312.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005312.hg.1 | TC06000507.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000474635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000474635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000474635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000474635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000474635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | ENST00000474635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000244520.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374017.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000374018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000474635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000474635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000474635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000474635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000474635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | ENST00000474635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007769.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153830.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153830.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153830.hg.1 | NM_003093.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153830.hg.1 | NR_029472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153830.hg.1 | TC0600007769.hg.1 | Clariom D TC |

SNRPC
Genomic Map