Assay Details
| Gene Symbol: FCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK075140.1 | - | 5 - 6 | 493 | 106 | |
| AK309540.1 | - | 6 - 7 | 522 | 106 | ||
| AY358081.1 | - | 5 - 6 | 482 | 106 | ||
| AY358283.1 | - | 5 - 6 | 519 | 106 | ||
| BC020731.1 | - | 5 - 6 | 493 | 4692272 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199763.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199763.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199763.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199763.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199763.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199763.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199763.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199763.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199763.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199763.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199763.hg.1 | TC0100013429.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |

FCN3
Genomic Map