Assay Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003083.3 | NP_003074.1 | 2 - 3 | 356 | 92 | |
| NR_030717.1 | 2 - 3 | 253 | 92 | |||
| GenBank mRNA | AK314878.1 | - | 2 - 3 | 341 | 92 | |
| BC011868.2 | - | 2 - 3 | 311 | 4545178 | 92 | |
| BP234322.1 | - | 2 - 3 | 252 | 92 | ||
| BX647416.1 | - | 2 - 3 | 220 | 92 | ||
| CF454397.1 | - | 2 - 3 | 356 | 92 | ||
| U44755.1 | - | 2 - 3 | 340 | 92 | ||
| U44898.1 | - | 2 - 3 | 322 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000142.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002251.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002251.hg.1 | TC19000142.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002253.hg.1 | ENST00000221573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002253.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002253.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002253.hg.1 | TC19000142.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148688.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148688.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148688.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148688.hg.1 | TC1900006866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900148688.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148688.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148688.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148686.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148686.hg.1 | TC1900006866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900148686.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | TC1900006866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148692.hg.1 | NR_030717.1 | Refseq |

SNAPC2
Genomic Map