Assay Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003083.3 | NP_003074.1 | 3 - 4 | 429 | 137 | |
| NR_030717.1 | 3 - 4 | 326 | 137 | |||
| GenBank mRNA | AK314878.1 | - | 3 - 4 | 414 | 137 | |
| BC011868.2 | - | 3 - 4 | 384 | 4545178 | 137 | |
| BP234322.1 | - | 3 - 4 | 325 | 138 | ||
| BX647416.1 | - | 3 - 4 | 293 | 137 | ||
| CF454397.1 | - | 3 - 4 | 429 | 137 | ||
| U44755.1 | - | 3 - 4 | 413 | 137 | ||
| U44898.1 | - | 3 - 4 | 395 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000142.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002254.hg.1 | ENST00000221573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002254.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002254.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002254.hg.1 | TC19000142.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | TC1900006866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148693.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | TC1900006866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900148694.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000221573.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000593598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000595637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000596520.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | ENST00000597584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NM_003083.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006866.hg.1 | NR_030717.1 | Refseq |

SNAPC2
Genomic Map