Assay Details
| Gene Symbol: FBXO28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136115.2 | NP_001129587.1 | 2 - 3 | 426 | 101 | |
| NM_015176.3 | NP_055991.1 | 2 - 3 | 426 | 101 | ||
| GenBank mRNA | AB007952.1 | - | 2 - 3 | 177 | 101 | |
| AK001628.1 | - | 2 - 3 | 402 | 101 | ||
| AK307892.1 | - | 2 - 3 | 398 | 101 | ||
| BC031576.1 | - | 2 - 3 | 395 | 5188219 | 101 | |
| BP249147.1 | - | 2 - 3 | 424 | 101 | ||
| DB136175.1 | - | 2 - 3 | 427 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001841.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000483773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000483773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000483773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000483773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000483773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000519894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000519894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000519894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028637.hg.1 | ENST00000366862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028637.hg.1 | ENST00000424254.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028637.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028637.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028637.hg.1 | TC01001841.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000483773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000483773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000483773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000483773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000483773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000519894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000519894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000519894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | ENST00000523990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011733.hg.1 | NR_049764.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186450.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186450.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186450.hg.1 | ENST00000483773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186450.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186450.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186450.hg.1 | ENST00000366862.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186450.hg.1 | ENST00000424254.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186450.hg.1 | ENST00000483773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186450.hg.1 | NM_001136115.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186450.hg.1 | NM_015176.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186450.hg.1 | TC0100011733.hg.1 | Clariom D TC |

FBXO28
Genomic Map