Assay Details
| Gene Symbol: SIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005069.4 | NP_005060.1 | 8 - 9 | 1608 | 67 | |
| NM_009586.3 | NP_033664.2 | 8 - 9 | 1608 | 67 | ||
| XM_011529694.1 | XP_011527996.1 | 7 - 8 | 1775 | 67 | ||
| GenBank mRNA | AJ001858.1 | - | 7 - 8 | 806 | 67 | |
| BC110444.2 | - | 8 - 9 | 1020 | 40007501 | 67 | |
| U80456.1 | - | 8 - 9 | 1086 | 67 | ||
| U80457.1 | - | 8 - 9 | 1086 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000157.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
nervous system development
embryonic pattern specification
cell differentiation
lung development
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein heterodimerization activity
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
nervous system development
embryonic pattern specification
cell differentiation
lung development
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21001384.hg.1 | ENST00000430056.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001384.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001384.hg.1 | TC21000157.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000460783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000460783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000460783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000460783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000460783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000483178.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000483178.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000483178.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | ENST00000483178.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001389.hg.1 | ENST00000290399.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001389.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001389.hg.1 | TC21000157.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2100147688.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147688.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147688.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147688.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147688.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147688.hg.1 | TC2100007056.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100147688.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147688.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147688.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100147688.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100147688.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000290399.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000431229.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000460783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000460783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000460783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000460783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000460783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000481185.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000483178.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000483178.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000483178.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | ENST00000483178.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_005069.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007056.hg.1 | NM_009586.4 | Refseq |

SIM2
Genomic Map