Assay Details
| Gene Symbol: GNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000172.3 | NP_000163.2 | 3 - 4 | 407 | 64 | |
| NM_144499.2 | NP_653082.1 | 3 - 4 | 407 | 64 | ||
| GenBank mRNA | AF493908.1 | - | 3 - 4 | 291 | 64 | |
| BC095505.1 | - | 3 - 4 | 294 | 6519160 | 64 | |
| BX953950.1 | - | 3 - 4 | 405 | 64 | ||
| X15088.1 | - | 3 - 4 | 394 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000305.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
detection of chemical stimulus involved in sensory perception of bitter taste
protein folding
signal transduction
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
visual perception
phototransduction, visible light
cell proliferation
response to light stimulus
response to light intensity
rhodopsin mediated signaling pathway
regulation of rhodopsin mediated signaling pathway
eye photoreceptor cell development
detection of light stimulus involved in visual perception
sensory perception of umami taste
positive regulation of cyclic-nucleotide phosphodiesterase activity
negative regulation of cyclic-nucleotide phosphodiesterase activity
retina development in camera-type eye
cellular response to electrical stimulus
acyl binding
G-protein coupled receptor binding
GTPase activity
signal transducer activity
GTP binding
GDP binding
protein kinase binding
G-protein beta/gamma-subunit complex binding
metal ion binding
detection of chemical stimulus involved in sensory perception of bitter taste
protein folding
signal transduction
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
visual perception
phototransduction, visible light
cell proliferation
response to light stimulus
response to light intensity
rhodopsin mediated signaling pathway
regulation of rhodopsin mediated signaling pathway
eye photoreceptor cell development
detection of light stimulus involved in visual perception
sensory perception of umami taste
positive regulation of cyclic-nucleotide phosphodiesterase activity
negative regulation of cyclic-nucleotide phosphodiesterase activity
retina development in camera-type eye
cellular response to electrical stimulus
acyl binding
G-protein coupled receptor binding
GTPase activity
signal transducer activity
GTP binding
GDP binding
protein kinase binding
G-protein beta/gamma-subunit complex binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03006417.hg.1 | ENST00000232461.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03006417.hg.1 | ENST00000433068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03006417.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03006417.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03006417.hg.1 | TC03000305.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03006418.hg.1 | ENST00000232461.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03006418.hg.1 | ENST00000433068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03006418.hg.1 | ENST00000481246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03006418.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03006418.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03006418.hg.1 | TC03000305.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000467787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000467787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000467787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000481246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000481246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000481246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | ENST00000481246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300152862.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152862.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152862.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152862.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300152862.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300152862.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152862.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152862.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152862.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300152862.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300152862.hg.1 | TC0300007408.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000467787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000467787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000467787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000481246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000481246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000481246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | ENST00000481246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007408.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300152863.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152863.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152863.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152863.hg.1 | ENST00000481246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152863.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300152863.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300152863.hg.1 | ENST00000232461.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152863.hg.1 | ENST00000433068.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152863.hg.1 | ENST00000440836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152863.hg.1 | ENST00000481246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152863.hg.1 | NM_000172.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300152863.hg.1 | NM_144499.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300152863.hg.1 | TC0300007408.hg.1 | Clariom D TC |

GNAT1
Genomic Map