Assay Details
| Gene Symbol: SURF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001280787.1 | NP_001267716.1 | 3 - 4 | 299 | 114 | |
| NM_003172.3 | NP_003163.1 | 4 - 5 | 351 | 114 | ||
| XM_011518942.1 | XP_011517244.1 | 3 - 4 | 401 | 114 | ||
| GenBank mRNA | AK291122.1 | - | 4 - 5 | 334 | 114 | |
| BC071658.1 | - | 4 - 5 | 356 | 5539506 | 114 | |
| BE905282.1 | - | 3 - 4 | 255 | 114 | ||
| KU178341.1 | - | 4 - 5 | 319 | 114 | ||
| KU178342.1 | - | 3 - 4 | 267 | 114 | ||
| KU178343.1 | - | 2 - 3 | 133 | 114 | ||
| Z35093.1 | - | 4 - 5 | 333 | 114 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001687.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001280787.1,XM_011518942.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000626556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000626556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021621.hg.1 | ENST00000371974.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021621.hg.1 | ENST00000495952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021621.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021621.hg.1 | TC09001687.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021623.hg.1 | ENST00000371974.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021623.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021623.hg.1 | TC09001687.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000626556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000626556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181058.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181058.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181058.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181058.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181058.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181058.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181058.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181058.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181058.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181058.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181058.hg.1 | TC0900011841.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900181055.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181055.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181055.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181055.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181055.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181055.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181055.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181055.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181055.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181055.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181055.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181055.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181055.hg.1 | TC0900011841.hg.1 | Clariom D TC |

SURF1
Genomic Map