Assay Details
| Gene Symbol: TEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024683.3 | NP_078959.3 | 2 - 3 | 562 | 92 | |
| XM_006722084.1 | XP_006722147.1 | 2 - 3 | 1046 | 92 | ||
| GenBank mRNA | AJ314645.1 | - | 2 - 3 | 529 | 92 | |
| BC009025.2 | - | 1 - 2 | 462 | 92 | ||
| BC024328.1 | - | 2 - 3 | 529 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001337.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000541382.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000541382.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000541382.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000541382.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000579183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000579183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000579183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017818.hg.1 | ENST00000306049.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017818.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017818.hg.1 | TC17001337.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017819.hg.1 | ENST00000306049.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017819.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017819.hg.1 | TC17001337.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000541382.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000541382.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000541382.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000541382.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000579183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000579183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000579183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010303.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180989.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180989.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180989.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180989.hg.1 | TC1700010303.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700180989.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180989.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180989.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180988.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180988.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180988.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180988.hg.1 | NM_024683.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700180988.hg.1 | TC1700010303.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700180988.hg.1 | ENST00000306049.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180988.hg.1 | ENST00000580840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180988.hg.1 | ENST00000581216.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700180988.hg.1 | NM_024683.3 | Refseq |

TEFM
Genomic Map