Assay Details
| Gene Symbol: NIPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144599.4 | NP_653200.2 | 1 - 2 | 200 | 72 | |
| GenBank mRNA | AK057846.1 | - | 1 - 2 | 55 | 72 | |
| AK314073.1 | - | 1 - 2 | 161 | 72 | ||
| BC103705.1 | - | 1 - 2 | 65 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15002770.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK314073.1,BC103705.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009596.hg.1 | ENST00000337435.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009596.hg.1 | ENST00000538684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009596.hg.1 | ENST00000561183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009596.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009596.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009596.hg.1 | TC15002770.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009598.hg.1 | ENST00000337435.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009598.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009598.hg.1 | TC15002770.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009597.hg.1 | ENST00000337435.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009597.hg.1 | ENST00000538684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009597.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009597.hg.1 | TC15002770.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500180785.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180785.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500180785.hg.1 | TC1500010691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500180785.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500180785.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180787.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180787.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180787.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180787.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500180787.hg.1 | TC1500010691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500180787.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180787.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180787.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180787.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | TC1500010691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500180789.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500180784.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180784.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500180784.hg.1 | TC1500010691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500180784.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180784.hg.1 | NM_144599.4 | Refseq |

NIPA1
Genomic Map