Assay Details
| Gene Symbol: NIPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142275.1 | NP_001135747.1 | 3 - 4 | 168 | 71 | |
| NM_144599.4 | NP_653200.2 | 3 - 4 | 347 | 71 | ||
| GenBank mRNA | AK057846.1 | - | 3 - 4 | 202 | 71 | |
| AK314073.1 | - | 3 - 4 | 308 | 71 | ||
| BC025678.1 | - | 3 - 4 | 168 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15002770.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15009593.hg.1 | ENST00000337435.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009593.hg.1 | ENST00000538684.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009593.hg.1 | ENST00000561183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009593.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009593.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15009593.hg.1 | TC15002770.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | TC1500010691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500180796.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000337435.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000557930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000559448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000560105.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | ENST00000561183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_001142275.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010691.hg.1 | NM_144599.4 | Refseq |

NIPA1
Genomic Map