Assay Details
| Gene Symbol: DOLPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020438.4 | NP_065171.2 | 5 - 6 | 490 | 59 | |
| GenBank mRNA | AK290713.1 | - | 5 - 6 | 490 | 59 | |
| AY189675.1 | - | 5 - 6 | 493 | 59 | ||
| BC009493.2 | - | 5 - 6 | 460 | 59 | ||
| BC033686.1 | - | 5 - 6 | 467 | 5195127 | 59 | |
| DA590931.1 | - | 5 - 6 | 490 | 59 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000707.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000412363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000412363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000412363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000412363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000412363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000412363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000480255.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000480255.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000480255.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | ENST00000480255.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008885.hg.1 | ENST00000372546.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008885.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09008885.hg.1 | TC09000707.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0900160168.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900160168.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900160168.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900160168.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900160168.hg.1 | TC0900008899.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000327812.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000372546.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000406974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000412363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000412363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000412363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000412363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000412363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000412363.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000480255.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000480255.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000480255.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | ENST00000480255.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_001135917.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NM_020438.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NR_072983.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008899.hg.1 | NR_072983.1 | Refseq |

DOLPP1
Genomic Map