Assay Details
| Gene Symbol: MTDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178812.3 | NP_848927.2 | 4 - 5 | 1073 | 144 | |
| XM_005251099.3 | XP_005251156.1 | 4 - 5 | 1280 | 144 | ||
| XM_011517367.2 | XP_011515669.1 | 4 - 5 | 1280 | 144 | ||
| XM_011517369.2 | XP_011515671.1 | 4 - 5 | 1280 | 144 | ||
| XM_011517370.2 | XP_011515672.1 | 4 - 5 | 1280 | 144 | ||
| XM_017013968.1 | XP_016869457.1 | 4 - 5 | 1280 | 144 | ||
| GenBank mRNA | AF411226.1 | - | 4 - 5 | 964 | 144 | |
| AF501310.1 | - | 4 - 5 | 1004 | 144 | ||
| AK000745.1 | - | 4 - 5 | 459 | 144 | ||
| AY082966.1 | - | 4 - 5 | 823 | 144 | ||
| AY974040.1 | - | 4 - 5 | 815 | 144 | ||
| BC001815.1 | - | 5 - 6 | 1013 | 144 | ||
| BC009324.2 | - | 5 - 6 | 928 | 144 | ||
| BC014977.1 | - | 4 - 5 | 1045 | 144 | ||
| BC045642.1 | - | 4 - 5 | 1073 | 5298467 | 144 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000594.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
positive regulation of autophagy
lipopolysaccharide-mediated signaling pathway
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of angiogenesis
positive regulation of NF-kappaB transcription factor activity
positive regulation of protein kinase B signaling
bicellular tight junction assembly
RNA polymerase II transcription factor binding
transcription coactivator activity
double-stranded RNA binding
protein binding
poly(A) RNA binding
NF-kappaB binding
negative regulation of transcription from RNA polymerase II promoter
positive regulation of autophagy
lipopolysaccharide-mediated signaling pathway
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of angiogenesis
positive regulation of NF-kappaB transcription factor activity
positive regulation of protein kinase B signaling
bicellular tight junction assembly
RNA polymerase II transcription factor binding
transcription coactivator activity
double-stranded RNA binding
protein binding
poly(A) RNA binding
NF-kappaB binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008226.hg.1 | ENST00000336273.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008226.hg.1 | ENST00000519934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008226.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008226.hg.1 | TC08000594.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008225.hg.1 | ENST00000336273.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008225.hg.1 | ENST00000519934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008225.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08008225.hg.1 | TC08000594.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0800157250.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157250.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157250.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157250.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157250.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157250.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157250.hg.1 | TC0800008314.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800157249.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157249.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157249.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157249.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157249.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157249.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157249.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800157249.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800157249.hg.1 | TC0800008314.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000336273.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000519934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000521933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | ENST00000522313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008314.hg.1 | NM_178812.3 | Refseq |

MTDH
Genomic Map