Assay Details
| Gene Symbol: FAM129A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052966.3 | NP_443198.1 | 12 - 13 | 1780 | 80 | |
| XM_011509140.2 | XP_011507442.1 | 13 - 14 | 2655 | 80 | ||
| XM_011509141.2 | XP_011507443.1 | 12 - 13 | 2544 | 80 | ||
| GenBank mRNA | AB050477.1 | - | 12 - 13 | 1706 | 80 | |
| AF288391.1 | - | 12 - 13 | 1744 | 80 | ||
| AK022527.1 | - | 12 - 13 | 1618 | 80 | ||
| AK095547.1 | - | 7 - 8 | 1024 | 80 | ||
| AY550972.1 | - | 12 - 13 | 1619 | 80 | ||
| BC030531.1 | - | 12 - 13 | 1636 | 5212338 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003624.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01055743.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055743.hg.1 | ENST00000487074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055743.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055743.hg.1 | TC01003624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000496299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000496299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226525.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226525.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226525.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226525.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226525.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226525.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226525.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226525.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226525.hg.1 | TC0100016678.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000496299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000496299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |

FAM129A
Genomic Map