Assay Details
| Gene Symbol: FAM129A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052966.3 | NP_443198.1 | 1 - 2 | 284 | 74 | |
| GenBank mRNA | AB050477.1 | - | 1 - 2 | 210 | 74 | |
| AF288391.1 | - | 1 - 2 | 248 | 74 | ||
| AK022527.1 | - | 1 - 2 | 122 | 74 | ||
| AK310557.1 | - | 1 - 2 | 151 | 74 | ||
| AK311019.1 | - | 1 - 2 | 151 | 74 | ||
| AY550972.1 | - | 1 - 2 | 123 | 74 | ||
| BC030531.1 | - | 1 - 2 | 140 | 5212338 | 74 | |
| DC346827.1 | - | 1 - 2 | 284 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003624.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB050477.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01055766.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055766.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055766.hg.1 | TC01003624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055764.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055764.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055764.hg.1 | TC01003624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000496299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000496299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055765.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055765.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055765.hg.1 | TC01003624.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100226546.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226546.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226546.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226546.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226546.hg.1 | TC0100016678.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000496299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000496299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226547.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226547.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226547.hg.1 | TC0100016678.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100226544.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226544.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226544.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226544.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226544.hg.1 | TC0100016678.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100226545.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226545.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226545.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226545.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226545.hg.1 | TC0100016678.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100226543.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226543.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226543.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226543.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226543.hg.1 | TC0100016678.hg.1 | Clariom D TC |

FAM129A
Genomic Map