Assay Details
| Gene Symbol: SPATA19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291992.1 | NP_001278921.1 | 1 - 2 | 172 | 129 | |
| NM_174927.2 | NP_777587.1 | 1 - 2 | 172 | 129 | ||
| GenBank mRNA | AF521713.1 | - | 1 - 2 | 108 | 129 | |
| AK098717.1 | - | 1 - 2 | 131 | 129 | ||
| BC058039.1 | - | 1 - 2 | 139 | 6619434 | 129 | |
| BX096445.1 | - | 1 - 2 | 172 | 129 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002468.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF521713.1,AK098717.1,BC058039.1,NM_001291992.1,NM_174927.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11029576.hg.1 | ENST00000299140.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029576.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029576.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029576.hg.1 | TC11002468.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029578.hg.1 | ENST00000299140.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029578.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029578.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029578.hg.1 | TC11002468.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029577.hg.1 | ENST00000299140.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029577.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029577.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11029577.hg.1 | TC11002468.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100198845.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198845.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198845.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198845.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198845.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198845.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198845.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198845.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198845.hg.1 | TC1100012902.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100198846.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198846.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198846.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198846.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198846.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198846.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198846.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198846.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198846.hg.1 | TC1100012902.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100198848.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198848.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198848.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198848.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198848.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198848.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198848.hg.1 | TC1100012902.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100198844.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198844.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198844.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198844.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198844.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198844.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198844.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198844.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198844.hg.1 | TC1100012902.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012902.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198847.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198847.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198847.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198847.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198847.hg.1 | ENST00000299140.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198847.hg.1 | ENST00000532889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100198847.hg.1 | NM_001291992.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198847.hg.1 | NM_174927.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100198847.hg.1 | TC1100012902.hg.1 | Clariom D TC |

SPATA19
Genomic Map