Assay Details
| Gene Symbol: PPP1R16B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172735.2 | NP_001166206.1 | 2 - 3 | 449 | 123 | |
| NM_015568.3 | NP_056383.1 | 2 - 3 | 449 | 123 | ||
| XM_017027785.1 | XP_016883274.1 | 2 - 3 | 448 | 123 | ||
| GenBank mRNA | AB020630.2 | - | 2 - 3 | 437 | 123 | |
| AB177855.1 | - | 1 - 2 | 76 | 123 | ||
| AF362910.1 | - | 1 - 2 | 300 | 123 | ||
| BC131801.1 | - | 2 - 3 | 406 | 40081818 | 123 | |
| BC152467.1 | - | 2 - 3 | 437 | 8862603 | 123 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000291.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017027785.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of endothelial cell proliferation
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of protein dephosphorylation
negative regulation of protein dephosphorylation
regulation of catalytic activity
regulation of filopodium assembly
establishment of endothelial barrier
negative regulation of peptidyl-serine dephosphorylation
positive regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
protein binding
protein phosphatase type 1 regulator activity
positive regulation of endothelial cell proliferation
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of protein dephosphorylation
negative regulation of protein dephosphorylation
regulation of catalytic activity
regulation of filopodium assembly
establishment of endothelial barrier
negative regulation of peptidyl-serine dephosphorylation
positive regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
protein binding
protein phosphatase type 1 regulator activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20004183.hg.1 | ENST00000299824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004183.hg.1 | ENST00000373331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004183.hg.1 | ENST00000468265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004183.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004183.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004183.hg.1 | TC20000291.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004186.hg.1 | ENST00000299824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004186.hg.1 | ENST00000373331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004186.hg.1 | ENST00000468265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004186.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004186.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004186.hg.1 | TC20000291.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150795.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150795.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150795.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150795.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150795.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150795.hg.1 | TC2000007336.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150795.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150795.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150795.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150795.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150795.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150798.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150798.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150798.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150798.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150798.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150798.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150798.hg.1 | TC2000007336.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150798.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150798.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150798.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150798.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150798.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150798.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |

PPP1R16B
Genomic Map