Assay Details
| Gene Symbol: PPP1R16B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172735.2 | NP_001166206.1 | 8 - 9 | 1101 | 122 | |
| NM_015568.3 | NP_056383.1 | 9 - 10 | 1227 | 122 | ||
| XM_011528768.2 | XP_011527070.1 | 8 - 9 | 1790 | 122 | ||
| XM_011528769.2 | XP_011527071.1 | 7 - 8 | 808 | 122 | ||
| XM_017027785.1 | XP_016883274.1 | 8 - 9 | 1122 | 122 | ||
| GenBank mRNA | AB020630.2 | - | 9 - 10 | 1215 | 122 | |
| AB177855.1 | - | 7 - 8 | 728 | 122 | ||
| AF362910.1 | - | 8 - 9 | 1078 | 122 | ||
| BC131801.1 | - | 9 - 10 | 1184 | 40081818 | 122 | |
| BC152467.1 | - | 9 - 10 | 1215 | 8862603 | 122 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000291.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of endothelial cell proliferation
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of protein dephosphorylation
negative regulation of protein dephosphorylation
regulation of catalytic activity
regulation of filopodium assembly
establishment of endothelial barrier
negative regulation of peptidyl-serine dephosphorylation
positive regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
protein binding
protein phosphatase type 1 regulator activity
positive regulation of endothelial cell proliferation
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of protein dephosphorylation
negative regulation of protein dephosphorylation
regulation of catalytic activity
regulation of filopodium assembly
establishment of endothelial barrier
negative regulation of peptidyl-serine dephosphorylation
positive regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
protein binding
protein phosphatase type 1 regulator activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20004195.hg.1 | ENST00000299824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004195.hg.1 | ENST00000373331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004195.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004195.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004195.hg.1 | TC20000291.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150807.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150807.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150807.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150807.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150807.hg.1 | TC2000007336.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150807.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150807.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150807.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150807.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000299824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000373331.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000463749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | ENST00000468265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_001172735.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007336.hg.1 | NM_015568.3 | Refseq |

PPP1R16B
Genomic Map