| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011903.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011903.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011903.hg.1 |
TC20000871.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000164391.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164391.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164391.hg.1 |
TC2000009204.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000164391.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164391.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
ENST00000372813.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009204.hg.1 |
NM_006809.4 |
Refseq |