Assay Details
| Gene Symbol: MCEE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032601.3 | NP_115990.3 | 2 - 3 | 427 | 94 | |
| GenBank mRNA | AF364547.1 | - | 2 - 3 | 400 | 94 | |
| BC020825.1 | - | 2 - 3 | 392 | 4734422 | 94 | |
| BM271757.1 | - | 2 - 3 | 449 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001973.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02031428.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031428.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031428.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031428.hg.1 | TC02001973.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031435.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031435.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031435.hg.1 | TC02001973.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031427.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031427.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031427.hg.1 | TC02001973.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000486135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000486135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000486135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000486135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000486135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195070.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195070.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195070.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195070.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195070.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195070.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195070.hg.1 | TC0200013019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000486135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000486135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000486135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000486135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000486135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013019.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195077.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195077.hg.1 | ENST00000486135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195077.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195077.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195077.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195077.hg.1 | ENST00000486135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195077.hg.1 | ENST00000494660.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195077.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195077.hg.1 | TC0200013019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200195072.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195072.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195072.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195072.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195072.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195072.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195072.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195072.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195072.hg.1 | TC0200013019.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200195071.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195071.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195071.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195071.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195071.hg.1 | ENST00000244217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195071.hg.1 | ENST00000413592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195071.hg.1 | ENST00000462609.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195071.hg.1 | NM_032601.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195071.hg.1 | TC0200013019.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 207809 | - | - |

MCEE
Genomic Map