| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003694.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003694.hg.1 |
ENST00000423219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003694.hg.1 |
ENST00000448498.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003694.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03003694.hg.1 |
TC03000190.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000466887.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
ENST00000496724.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300007074.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149500.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149500.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149500.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149500.hg.1 |
ENST00000273173.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149500.hg.1 |
ENST00000448498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300149500.hg.1 |
NM_004803.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300149500.hg.1 |
TC0300007074.hg.1 |
Clariom D TC |