Assay Details
| Gene Symbol: MTDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011517368.2 | XP_011515670.1 | 4 - 5 | 1285 | 77 | |
| XM_017013966.1 | XP_016869455.1 | 4 - 5 | 1285 | 77 | ||
| XM_017013967.1 | XP_016869456.1 | 4 - 5 | 1285 | 77 | ||
More Information
Set Membership: |
|

MTDH
Genomic Map