| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004041.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004041.hg.1 |
ENST00000462448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004041.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004041.hg.1 |
TC07000266.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004040.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004040.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004040.hg.1 |
TC07000266.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000462448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000462448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000462448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000462448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000474211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000474211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000474211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000474211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000474211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
ENST00000474211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004038.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004038.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004038.hg.1 |
TC07000266.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700150909.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150909.hg.1 |
ENST00000462448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150909.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150909.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150909.hg.1 |
ENST00000462448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150909.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150909.hg.1 |
TC0700007345.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000462448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000462448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000462448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000462448.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000474211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000474211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000474211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000474211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000474211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
ENST00000474211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007345.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150908.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150908.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150908.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150908.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150908.hg.1 |
TC0700007345.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700150907.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150907.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150907.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150907.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150907.hg.1 |
TC0700007345.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700150905.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150905.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150905.hg.1 |
ENST00000319357.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150905.hg.1 |
NM_004760.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150905.hg.1 |
TC0700007345.hg.1 |
Clariom D TC |