Assay Details
| Gene Symbol: SGPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003901.3 | NP_003892.2 | 11 - 12 | 1284 | 127 | |
| XM_005270263.1 | XP_005270320.1 | 11 - 12 | 1297 | 127 | ||
| XM_006718053.1 | XP_006718116.1 | 12 - 13 | 1399 | 127 | ||
| XM_011540316.2 | XP_011538618.1 | 11 - 12 | 1343 | 127 | ||
| XM_011540317.1 | XP_011538619.1 | 11 - 12 | 1193 | 127 | ||
| XM_011540318.1 | XP_011538620.1 | 10 - 11 | 1214 | 127 | ||
| GenBank mRNA | AB033078.1 | - | 11 - 12 | 1262 | 127 | |
| AF144638.1 | - | 10 - 11 | 1062 | 127 | ||
| AJ011304.2 | - | 11 - 12 | 1239 | 127 | ||
| AK308610.1 | - | 4 - 5 | 597 | 127 | ||
| AK314615.1 | - | 11 - 12 | 1193 | 127 | ||
| BC052991.1 | - | 11 - 12 | 1272 | 6150776 | 127 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000434.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
luteinization
vasculogenesis
kidney development
fatty acid metabolic process
ceramide metabolic process
spermatogenesis
androgen metabolic process
estrogen metabolic process
post-embryonic development
fibroblast migration
hemopoiesis
sphingolipid biosynthetic process
sphingolipid catabolic process
Leydig cell differentiation
regulation of multicellular organism growth
platelet-derived growth factor receptor signaling pathway
skeletal system morphogenesis
palate development
face morphogenesis
apoptotic signaling pathway
protein binding
sphinganine-1-phosphate aldolase activity
carboxy-lyase activity
pyridoxal phosphate binding
luteinization
vasculogenesis
kidney development
fatty acid metabolic process
ceramide metabolic process
spermatogenesis
androgen metabolic process
estrogen metabolic process
post-embryonic development
fibroblast migration
hemopoiesis
sphingolipid biosynthetic process
sphingolipid catabolic process
Leydig cell differentiation
regulation of multicellular organism growth
platelet-derived growth factor receptor signaling pathway
skeletal system morphogenesis
palate development
face morphogenesis
apoptotic signaling pathway
protein binding
sphinganine-1-phosphate aldolase activity
carboxy-lyase activity
pyridoxal phosphate binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10005102.hg.1 | ENST00000373202.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005102.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005102.hg.1 | TC10000434.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000299297.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000299297.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000299297.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000299297.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000299297.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000299297.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000409118.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000409118.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000409118.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000409118.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000486993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000486993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000486993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000486993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000611290.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000611290.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000611290.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000611290.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000613857.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000613857.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000613857.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000613857.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000613857.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000613857.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153883.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153883.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153883.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153883.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153883.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153883.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153883.hg.1 | TC1000007936.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000299297.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000299297.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000299297.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000299297.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000299297.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000299297.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000409118.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000409118.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000409118.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000409118.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000486993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000486993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000486993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000486993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000611290.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000611290.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000611290.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000611290.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000613857.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000613857.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000613857.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000613857.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000613857.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000613857.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000618021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | ENST00000620724.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007936.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153885.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153885.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153885.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153885.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153885.hg.1 | TC1000007936.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000153884.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153884.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153884.hg.1 | ENST00000373202.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153884.hg.1 | NM_003901.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153884.hg.1 | TC1000007936.hg.1 | Clariom D TC |

SGPL1
Genomic Map